Learn More
MP Biomedicals™ 1/4 in. Ceramic Spheres
Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Spheres include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
Brand:  MP Biomedicals™ 116540424
156.26 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 10548285
  • £180.50 / Pack of 100
Estimated Shipment: 02-08-2024
to see stock.
Description
Description
  • Zirconium Oxide-coated ceramic grinding spheres
  • Chemically inert
  • Component of Lysing Matrix A and M tubes
Specifications
Specifications
Zirconium oxide coated ceramic
Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
Bulk Matrix
Unlimited
Room Temperature
Break hard/brittle samples
100 Pk.
1/4 in. Zirconium Oxide Banded Satellite for Grinding
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.