Learn More
MP Biomedicals™ 1/4 in. Ceramic Spheres
Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Spheres include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
Brand: MP Biomedicals™ 116540034
60.72 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.05000kg
Description
- Zirconium Oxide-coated ceramic grinding spheres
- Chemically inert
- Component of Lysing Matrix A and M tubes
Specifications
Zirconium oxide coated ceramic | |
Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores | |
Bulk Matrix | |
Unlimited |
Room Temperature | |
Break hard/brittle samples | |
50 Pk. | |
1/4 in. Zirconium Oxide Banded Satellite for Grinding |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.