missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ (Arg8)-Vasopressin

Brand:  MP Biomedicals™ 0215296205


 View more versions of this product

Product Code. 11454091

  • £497.00 / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contains a disulfide bond between Cys1 and Cys6

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Biology and Analysis
(Arg-8)-Vasopressin (AVP)
-20°C
1084.23
5 mg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • [Arg8]-Vasopressin

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H332-Harmful if inhaled.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : [Arg8]-Vasopressin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers