missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ (Arg8)-Vasopressin
£491.84
Specifications
For Use With (Application) | Cell Biology and Analysis |
---|---|
Molecular Weight (g/mol) | 1084.23 |
Name | (Arg-8)-Vasopressin (AVP) |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11454091
|
MP Biomedicals™
0215296205 |
5mg |
£491.84
5mg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Contains a disulfide bond between Cys1 and Cys6
Specifications
Cell Biology and Analysis | |
1084.23 | |
(Arg-8)-Vasopressin (AVP) | |
-20°C |