missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ BigPrep™ Adapter for 2 x 50 mL Tube Holder on FastPrep-24

BigPrep™ Adapter for 2 x 50 mL Tube Holder on FastPrep-24 is used to grind or homogenize samples at ambient temperature.

Brand:  MP Biomedicals™ 116002525

1314.54 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11472380

  • £1495.00 / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Product Overview: BigPrep™ 2 x 50 mL Adapter is a large-scale adapter that holds 2 x 50 mL Lysing Matrix tubes (compatible with SafTest™ Food Inspection System). A wide range of disposable 50 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds while avoiding cross contamination occurring with traditional homogenization methods such as syringe shearing, sonication, or mortar and pestle grinding.
  • Applications: BigPrep™ 2 x 50 mL Adapter is a large sample volume adapter that is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Capacity: 2 tubes
  • Sample Volume: Up to 40 g
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Up to 40 g
Ideal for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing and forensic applications
FastPrep™- 24, FastPrep™-96
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119

    Expires2025-04-30

    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ BigPrep™ Adapter for 2 x 50 mL Tube Holder on FastPrep-24 > BigPrep™ Adapter for 2 x 50 mL Tube Holder on FastPrep-24

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.