missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Biopulverizer™ Lysing Matrix Set II

Biopulverizer™ Lysing Matrix Set II contains 10 x 2 mL tubes of each of specialized Lysing Matrix types (F, G, H, I and J).

Brand:  MP Biomedicals™ 116850200

Product Code. 11412410

  • £214.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes:
  • Matrix F: Aluminum Oxide, Alpha Alumina Crystals; Silicon Carbide, Hemihedral Crystals
  • Matrix G: Silicon Carbide, Hemihedral Crystals; Glass Beads, Spherical
  • Matrix H: Glass Beads, Spherical; Yellow Zirconium Oxide Beads, Spherical
  • Matrix I: Yellow Zirconium Oxide Beads, Spherical; 4 mm ceramic bead, Spherical
  • Matrix J: Yellow Zirconium Oxide Beads, Spherical; Aluminum Oxide, Alpha Alumina Crystals
This item is not returnable. View return policy

Description

Description

  • The Biopulverizer Set II contains 10 x 2 mL tubes of each of our specialized Lysing Matrix types F,G,H,I and J.
  • This set is ideal for optimizing homogenization conditions and protocols for very difficult, unusual or multiple samples.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nucleic Acid Isolation & Purification
50 Tubes
Suitable for all sample types
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Biopulverizer™ Lysing Matrix Set II > Biopulverizer™ Lysing Matrix Set II; Quantity: 50 Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.