missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Implen™ NanoPhotometer™ Standard Solutions
Use to control the photometric accuracy of the NanoPhotometer™ NP120, NP80, N60 and N50 (NanoVolume)
Brand: Implen™ N-568-S
Additional Details : Weight : 0.10000kg
Description
- Standard solutions for the control of photometric accuracy
- Verifies the instrument's absorbance accuracy
- For use with NanoPhotometer™ NP120/NP80/N60/N50
- CAS-No: 877-24-7
Specifications
Non-toxic Potassium Hydrogen Phthalate Solution (PHP) | |
10 x 0.5 mL | |
Store at room temperature |
NanoPhotometer N60 and NP80 | |
10 Vials | |
280 nm |
For Research Use Only