missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Brewers Yeast

Used as an insect diet.

Brand:  MP Biomedicals™ 0290331280

 View more versions of this product

Product Code. 11435722

  • £775.00 / 100lb

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Source of B-complex vitamins, chromium and selenium
  • Brewers Yeast is commonly used for baking break, fermenting wine and for beer and ethanol production
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Animal Diet
Source of B-complex Vitamins, Chromium and Selenium, also used to bake bread, ferment wine, beer and Ethanol production
S. cerevisiae
Powder
45.35 kg
15°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Brewers Yeast > Brewers Yeast, 100 lbs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.