missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Dextrose
Monohydrate
£154.76
Specifications
Product Type | Animal Diet |
---|---|
Format | Powder |
For Use With (Application) | Used as a supplement for cell culture and in numerous cellular processes |
Storage Requirements | 15°C to 30°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11422781
|
MP Biomedicals™
0290559425 |
11.33 kg |
£154.76
25lb |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Dextrose monohydrate is a common natural sugar involved in processes such as energy production, glycosylation and formation of glycans that provide structure to cells. It is involved in a detrimental process in cells called glycation. It is used as a supplement for cell culture and in numerous cellular processes.
Formula: C6H12O6•H2O
Formula Weight: 198.18
CAS No.: 14431-43-7
Specifications
Animal Diet | |
Powder | |
Used as a supplement for cell culture and in numerous cellular processes | |
15°C to 30°C |