missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ FastPROTEIN™ BlueMatrix
FastPROTEIN™ BlueMatrix Kit designed for the rapid extraction of proteins from gram (+/-) bacteria in a few seconds.
£227.90 - £354.04
Specifications
Product Type | Blue matrix kit |
---|---|
Includes | Matrix B, 0.1 mm Zirconium Silicate, Tubes |
Content And Storage | 15°C to 30°C |
Capacity | 2 mL |
For Use With (Application) | Rapid isolation and purification of Protein from bacterial cultures |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11452400
|
MP Biomedicals™
116550400 |
50 Preps |
£227.90
Pack of 50 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
11462400
|
MP Biomedicals™
116550500 |
100 Preps |
£354.04
Pack of 100 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
Description
- Kit or Assay Type: Protein Isolation, Mini-prep
- Molecule to be Enriched: Proteins
- Sample Type: Bacteria, Gram +/-
- Tube Size: 2 mL
- Matrix Composition: Matrix B, 0.1 mm zirconium silicate
Specifications
Blue matrix kit | |
15°C to 30°C | |
Rapid isolation and purification of Protein from bacterial cultures |
Matrix B, 0.1 mm Zirconium Silicate, Tubes | |
2 mL | |
FastProtein Blue Matrix |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.