Learn More
MP Biomedicals™ Lysing Matrix G
Lysing Matrix G Tubes for optimal lysis used with FastPrep™ instruments.
Brand: MP Biomedicals™ 116916050
205.43 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 80
Additional Details : Weight : 0.10000kg
Description
Silicon carbide has relatively low density and high strength and hardness, and therefore is effective at breaking tough, hard, or brittle cell membranes. Useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.
- Impact-resistant 2 mL tube contains 1.6 mm Silicon Carbide particles and 2 mm glass beads
- With brown caps
- Specific Gravity: Silicon Carbide: 3.20; Glass beads: 2.55
- Nucleic Acid Isolation and Purification
Specifications
2 mL | |
Dnase-free | |
Lysing matrix G | |
50 x 2 mL Tubes |
Spores, Animal Tissues, Fungi, Plants, Yeast | |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
Silicon Carbide, SiC; Glass, SiO2 | |
RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.