missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix G
Lysing Matrix G Tubes for optimal lysis used with FastPrep™ instruments.
£216.24 - £342.38
Specifications
Capacity | 2 mL |
---|---|
Sample Type | Spores, Animal Tissues, Fungi, Plants, Yeast |
DNase | Dnase-free |
For Use With (Application) | Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis |
Matrix | Lysing matrix G |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11402430
|
MP Biomedicals™
116916050 |
50 x 2 mL Tubes |
£216.24
Pack of 50 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11412430
|
MP Biomedicals™
116916100 |
100 x 2 mL Tubes |
£342.38
Pack of 100 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Silicon carbide has relatively low density and high strength and hardness, and therefore is effective at breaking tough, hard, or brittle cell membranes. Useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.
- Impact-resistant 2 mL tube contains 1.6 mm Silicon Carbide particles and 2 mm glass beads
- With brown caps
- Specific Gravity: Silicon Carbide: 3.20; Glass beads: 2.55
- Nucleic Acid Isolation and Purification
Specifications
2 mL | |
Dnase-free | |
Lysing matrix G |
Spores, Animal Tissues, Fungi, Plants, Yeast | |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.