Learn More
MP Biomedicals™ Lysing Matrix I
Lysing Matrix I is effective at breaking very tough and hard samples, including organisms with a tough or dense exterior matrix.
Brand: MP Biomedicals™ 116918050
205.02 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 80
Additional Details : Weight : 0.10000kg
Description
- Tube Size: 2 mL
- Sample Type: Plant and animal tissues, wood, seeds, very dense soils and clays, whole insects, ticks, ancient and dried samples, dry grinding fungal spores/rusts
- Molecule to be Enriched: DNA, RNA, proteins, other small molecules
- Matrix Composition: Matrix I, contains 2 mm yellow zirconia beads and a 4 mm black ceramic bead
Specifications
Seeds, Spores, Animal Tissues, Plants, Soil, Ancient and Dried Samples, Clay, Wood | |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
50 x 2 mL Tubes |
Dnase-free | |
Lysing matrix I | |
RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.