missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix I
Lysing Matrix I is effective at breaking very tough and hard samples, including organisms with a tough or dense exterior matrix.
Brand: MP Biomedicals™ 116918100
This item is not returnable.
View return policy
Description
- Tube Size: 2 mL
- Sample Type: Plant and animal tissues, wood, seeds, very dense soils and clays, whole insects, ticks, ancient and dried samples, dry grinding fungal spores/rusts
- Molecule to be Enriched: DNA, RNA, proteins, other small molecules
- Matrix Composition: Matrix I, contains 2 mm yellow zirconia beads and a 4 mm black ceramic bead
Specifications
Seeds, Spores, Animal Tissues, Plants, Soil, Ancient and Dried Samples, Clay, Wood | |
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
100 x 2 mL Tubes |
Dnase-free | |
Lysing matrix I | |
RNase-free |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction