Learn More
MP Biomedicals™ Plant Culture Salt Mixtures
Plant Culture Salt Mixtures are for the molecular testing of plants and plant-derived samples for improving the productivity, quality, and disease resistance of plants.
Brand: MP Biomedicals™ 092621820
53.54 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : CAS Number : 13477-34-4 Weight : 10.00000kg
Description
- Ready to use
- Liquid formulation, prepared according to the original specification, except where noted
- Available in different pack sizes, ready to use and liquid formulations
Specifications
Powder | |
Solution | |
10 x 1 L |
Hoagland's Modified Basal Salt Mixture | |
4.8 |