missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protein A, Fc Binding Grade

Fc Binding Grade, White Lyophilized Powder. Routinely used for immunoprecipitation assays.

Brand:  MP Biomedicals™ 0215389105

 View more versions of this product

Product Code. 11467342

  • £207.00 / 5mg
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Binding capacity 12-15mg of human lgG/mg solid
  • Carboxy truncated material containing 301 amino acid residues
  • White lyophilized powder
  • Source: Staphlococcus aureus , Cowan strain-1

TRUSTED_SUSTAINABILITY
Specifications

Specifications

12 to 15 mg of Human lgG/mg solid
-20°C
Lyophilized Powder
For detection of immunglobulins, purification
5 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.