missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protein A, Fc Binding Grade

Fc Binding Grade, White Lyophilized Powder. Routinely used for immunoprecipitation assays.

£58.90 - £207.00

Specifications

Binding Capacity 12 to 15 mg of Human lgG/mg solid
For Use With (Application) For detection of immunglobulins, purification
Content And Storage -20°C
Form Lyophilized Powder
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11457342
View Documents
MP Biomedicals™
0215389101
1 mg
£58.90
1mg
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
 
11467342
View Documents
MP Biomedicals™
0215389105
5 mg
£207.00
5mg
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
 
Description

Description

  • Binding capacity 12-15mg of human lgG/mg solid
  • Carboxy truncated material containing 301 amino acid residues
  • White lyophilized powder
  • Source: Staphlococcus aureus , Cowan strain-1

Specifications

Specifications

12 to 15 mg of Human lgG/mg solid
For detection of immunglobulins, purification
-20°C
Lyophilized Powder
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.