Learn More
MP Biomedicals™ Ready to Use 5% 7X™ Solutions
A 1 gallon Ready to Use 5% solution of 7X that includes a hand-pump for convenient dispensing
Brand: MP Biomedicals™ 097668493
Additional Details : Weight : 0.01000kg
Description
- Ideal for glass and plastic labware
- Contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware
- Environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds
Specifications
Liquid | |
15°C to 30°C | |
For Glass, Plastic, benchtop equipment and tubing |
7X | |
7X-O-Matic; 1 gal. | |
1 gal. |