missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ready to Use 5% 7X™ Solutions
A 1 gallon Ready to Use 5% solution of 7X that includes a hand-pump for convenient dispensing
£62.43 - £76.32
Specifications
Form | Liquid |
---|---|
Concentration | 7X |
Content And Storage | 15°C to 30°C |
For Use With (Application) | For Glass, Plastic, benchtop equipment and tubing |
Volume (English) | 1 gal. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11491495
|
MP Biomedicals™
097668193 |
ES 7X; 1 gal. |
£76.32
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11401505
|
MP Biomedicals™
097668493 |
7X-O-Matic; 1 gal. |
£62.43
Each |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Ideal for glass and plastic labware
- Contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware
- Environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds
Specifications
Liquid | |
15°C to 30°C | |
1 gal. |
7X | |
For Glass, Plastic, benchtop equipment and tubing |