missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ready to Use 5% 7X™ Solutions

A 1 gallon Ready to Use 5% solution of 7X that includes a hand-pump for convenient dispensing

Brand:  MP Biomedicals™ 097668193

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11491495

  • £76.32 / Each
Estimated Shipment: 15-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Ideal for glass and plastic labware
  • Contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware
  • Environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds

Specifications

Specifications

Liquid
15°C to 30°C
For Glass, Plastic, benchtop equipment and tubing
7X
ES 7X; 1 gal.
1 gal.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers