missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Wheat Germ Agglutinin
MP Biomedicals™ Wheat Germ Agglutinin has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers.
£268.18
Specifications
Color | White to Pale Yellow |
---|---|
Source | Triticum vulgaris |
Content And Storage | -20°C |
Form | Lyophilized |
For Use With (Application) | Proteomics, immune system regulation |
Description
- Less than 4 mg/mL will agglutinate human type O erythrocytes, and less than 1 mg/mL will agglutinate neuraminidase-treated erythrocytes
- Contains no protein-bound carbohydrate
Specifications
White to Pale Yellow | |
-20°C | |
Proteomics, immune system regulation |
Triticum vulgaris | |
Lyophilized | |
Lectin |