Learn More
MP Biomedicals™ MP Magnetic Rack 24
This tool is for fast and easy extraction of nucleic acids using a magnetic method. It is commonly used in combination with MagBeads FastDNA™ kits.
Brand: MP Biomedicals™ 116570413
552.86 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Magnet made of high-energy neodymium
- Small, strong and easy to handle
- For fast and efficient magnetic separations
- Design allows easy removal of rack from magnet
- Tubes can be placed on shaker or vortex during extraction process
- Good visibility of samples
- Up to 24 tubes of 1.5 or 2 mL
Storage:
- Magnet:
- Stored in a dry and cool place
- Never use > 50 °C
- Tube rack:
- Store at room temp
- Can be placed with tubes at temp as low as -20 °C for a limited time
Cleaning:
- Use with one of the following solutions:
- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution
Notes:
- Avoid prolonged exposure of rack to water or aqueous solutions
- Not autoclavable

Specifications
24 x 1.5 or 2 mL tubes | |
Store magnet in dry, cool place, Store rack at room temp | |
For use with the Magbeads FastDNA™ kit, tubes | |
Magnetic Rack |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.