missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14234-1-AP-150UL
This item is not returnable.
View return policy
Description
Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.Specifications
MPI | |
Polyclonal | |
Unconjugated | |
MPI | |
Mannose 6 phosphate isomerase, mannose phosphate isomerase, MPI, Phosphohexomutase, Phosphomannose isomerase, PMI, PMI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110119, 300741, 4351 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P34949, Q68FX1, Q924M7 | |
MPI | |
MPI Fusion Protein Ag5475 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |