Learn More
Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform
Improve hybrid MS bio-analysis with this exclusive workflow solution for proteins and peptides.
Brand: Thermo Scientific™ 991001024
Description
Improve hybrid MS bio-analysis with Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform , an exclusive Thermo Fisher Scientific workflow solution for proteins and peptides. Enabled through proprietary MSIA D.A.R.T.'S™ technologies, these workflows have demonstrated pg/mL level detection while providing simultaneous qualitative/quantitative data content from complex biological matrix, such as human plasma. Ideal for biologic and biomarker bio-analysis, MSIA provides innovative enablement in the areas of PKPD, DMPK, ADME, Biotransformation (DAR and payload determination), as well as routine biomarker analysis.
Next generation hybrid Mass Spectrometry bio-analytical workflows:
- Enables analysis of proteins of interest down to femtomole level
- Streamlined workflow for protein qualitative & quantative bio-analysis
- Employs a proprietary molecular trapping micro-column technology in an easy to use pipette tip housing format
- Drives antibody/antigen binding kinetics and greatly improves assaying of low-abundance protein from a biological sample
Enabling workflows and technologies for biologic and biomarker research:
- Immunoaffinity column captures protein of interest with improved signal-to-noise ratio
- Superior specificity and assaying characteristics compared to conventional approaches
- Allows for hybrid assaying down to femtomole levels
- Streptavidin and Protein A, G & A/G MSIA D.A.R.T.'S provide end user flexibility to analyze their specific targets of interest
- Custom MSIA D.A.R.T.'S are available upon request
- Optimized workflow solution Insulin (endogenous and therapeutic forms)
Requires:
300 μL pipette such as Thermo Scientific™ Versette™ Liquid Handling Platform, Thermo Scientific™ Finnpipette™ Novus i™ Multichannel pipetters (listed in user manual)
Recommended for:
Hybrid mass spectrometry based assays
For research use only
Specifications
MSIA D.A.R.T.'S, Insulin | |
300 μL | |
Pack of 24 Tips | |
MSIA D.A.R.T.S |