Learn More
Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform
Improve hybrid MS bio-analysis with this exclusive workflow solution for proteins and peptides.
£57.40 - £1640.00
Product Code | Brand | Description | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Format | Price | Quantity & Availability | ||||
15182579
|
Thermo Scientific™
991CUS02 |
MSIA D.A.R.T.'S, Custom | Pack of 96 Tips | N/A | |||||
15152579
|
Thermo Scientific™
991PRT11 |
MSIA D.A.R.T.'S, Protein A | Pack of 96 Tips | N/A | |||||
15153323
|
Thermo Scientific™
991PRT12 |
MSIA D.A.R.T.'S, Protein A | Pack of 24 Tips | N/A | |||||
15132579
|
Thermo Scientific™
991PRT13 |
MSIA D.A.R.T.'S, Protein G | Pack of 96 Tips | N/A | |||||
15142579
|
Thermo Scientific™
991PRT14 |
MSIA D.A.R.T.'S, Protein G | Pack of 24 Tips | N/A | |||||
15162579
|
Thermo Scientific™
991PRT15 |
MSIA D.A.R.T.'S, Protein A/G | Pack of 96 Tips | N/A | |||||
13288009
|
Thermo Scientific™
991PRT16 |
MSIA D.A.R.T.'S, Protein A/G | Pack of 24 Tips | N/A | |||||
15149628
|
Thermo Scientific™
991STR11 |
MSIA D.A.R.T.'S, Streptavidin | Pack of 96 Tips | N/A | |||||
15132589
|
Thermo Scientific™
991STR12 |
MSIA D.A.R.T.'S, Streptavidin | Pack of 24 Tips | N/A | |||||
15287608
|
Thermo Scientific™
991001096 |
MSIA D.A.R.T.'S, Insulin | Pack of 96 Tips | N/A | |||||
15247648
|
Thermo Scientific™
991001024 |
MSIA D.A.R.T.'S, Insulin | Pack of 24 Tips | N/A | |||||
15192579
|
Thermo Scientific™
991R |
MSIA D.A.R.T.'S, Reloadable Rack | Reload Rack (tips not included) | N/A | |||||
Description
Improve hybrid MS bio-analysis with Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform , an exclusive Thermo Fisher Scientific workflow solution for proteins and peptides. Enabled through proprietary MSIA D.A.R.T.'S™ technologies, these workflows have demonstrated pg/mL level detection while providing simultaneous qualitative/quantitative data content from complex biological matrix, such as human plasma. Ideal for biologic and biomarker bio-analysis, MSIA provides innovative enablement in the areas of PKPD, DMPK, ADME, Biotransformation (DAR and payload determination), as well as routine biomarker analysis.
Next generation hybrid Mass Spectrometry bio-analytical workflows:
- Enables analysis of proteins of interest down to femtomole level
- Streamlined workflow for protein qualitative & quantative bio-analysis
- Employs a proprietary molecular trapping micro-column technology in an easy to use pipette tip housing format
- Drives antibody/antigen binding kinetics and greatly improves assaying of low-abundance protein from a biological sample
Enabling workflows and technologies for biologic and biomarker research:
- Immunoaffinity column captures protein of interest with improved signal-to-noise ratio
- Superior specificity and assaying characteristics compared to conventional approaches
- Allows for hybrid assaying down to femtomole levels
- Streptavidin and Protein A, G & A/G MSIA D.A.R.T.'S provide end user flexibility to analyze their specific targets of interest
- Custom MSIA D.A.R.T.'S are available upon request
- Optimized workflow solution Insulin (endogenous and therapeutic forms)
Requires:
300 μL pipette such as Thermo Scientific™ Versette™ Liquid Handling Platform, Thermo Scientific™ Finnpipette™ Novus i Multichannel pipetters (listed in user manual)
Recommended for:
Hybrid mass spectrometry based assays
For research use only
Specifications
300 μL | |
MSIA D.A.R.T.S |