Learn More
Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1 Mouse Monoclonal Antibody, Epredia™
Deliver accurate, reproducible results with the Epredia™ Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1, Mouse Monoclonal Antibody.
£105.06 - £605.00
Specifications
Antigen | Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1 |
---|---|
Clone | 45M1 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12619866
|
Epredia
MS-145-B0 |
Concentrated | Biotin | 100 μL | |||||
12639866
|
Epredia
MS-145-B1 |
Concentrated | Biotin | 500 μL |
£487.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12629866
|
Epredia
MS-145-B |
Concentrated | Biotin | 1 mL |
£605.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12659866
|
Epredia
MS-145-P0 |
Concentrated | Unconjugated | 100 μL | |||||
12669866
|
Epredia
MS-145-P1ABX |
Concentrated | Unconjugated | 100 μL | |||||
12679866
|
Epredia
MS-145-PABX |
Concentrated | Unconjugated | 200 μL | |||||
11680443
|
Epredia
MS-145-P1 |
Concentrated | Unconjugated | 500 μL |
£311.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12649866
|
Medac
MS-145-P |
Concentrated | Unconjugated | 1 mL |
£600.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12609876
|
Epredia
MS-145-R7 |
Prediluted | Unconjugated | 7 mL | |||||
12619876
|
Epredia
MS-145-RQ |
Ready-to-Use | Unconjugated | 12 mL | |||||
Description
Mucins are high molecular eight glycoproteins with 80% carbohydrate contents and the remaining 20% is constituted by protein core. Gastric mucin is found in normal human gastrointestinal tract. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.Host Species: Mouse
Clone: 45M1
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Rabbit, Cat, Mouse, Rat, Pig, Hedgehog and Chicken. Does not react with cow. Others not known.
Epitope: Destroyed by β-mercaptoethanol and proteases but not by periodate treatment
Immunogen: M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient
Molecular Weight: >1000kDa
Positive Control: Stomach
Cellular Localization: Cytoplasmic and cell surface
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1 | |
Immunohistochemistry (Paraffin) | |
Mouse | |
Cancer and Tumor Biology | |
Primary |
45M1 | |
Monoclonal | |
IVD | |
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient |