Learn More
Nabertherm™ Muffle Furnace With Lift Door, 1100°C
Muffle furnaces are the reliable and long-lasting all-rounders in the laboratory and are ideally suited for a large number of processes in the field of material research and heat treatment.
£3380.00 - £7300.00
Specifications
Temperature (Metric) 1100°C
Type Furnace
Style Lift Door
Frequency 50/60 Hz
View More Specs
Products 12
Product Code Brand Capacity (Metric) Controller Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Controller Type Price Quantity & Availability  
17347849
View Documents
Nabertherm™
L-035H2ANL
3 L B510
£3380.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17357849
View Documents
Nabertherm™
L-035H2ANO
3 L C550
£4045.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17327859
View Documents
Nabertherm™
L-055H2ANL
5 L B510
£4065.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17337859
View Documents
Nabertherm™
L-055H2ANO
5 L C550
£4740.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17387869
View Documents
Nabertherm™
L-095H2ANL
9 L B510
£4690.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17397869
View Documents
Nabertherm™
L-095H2ANO
9 L C550
£5355.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17347889
View Documents
Nabertherm™
L-155H2ANL
15 L B510
£4905.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17357889
View Documents
Nabertherm™
L-155H2ANO
15 L C550
£5575.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17307909
View Documents
Nabertherm™
L-245H2ANL
24 L B510
£6620.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17317909
View Documents
Nabertherm™
L-245H2ANO
24 L C550
£7300.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17387909
View Documents
Nabertherm™
L-405H2ANL
40 L B510
£6940.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
17397909
View Documents
Nabertherm™
L-405K1ANL
40 L C550
£6030.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
Description
Description
  • Optimal temperature uniformity of +/- 5K with closed fresh air inlet in empty workspace
  • Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Ceramic heating plates with integral heating elements which are safeguarded and easy to replace
  • Adjustable air inlet integrated in door
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low-noise operation
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
  • Defined application within the constraints of the operating instructions
  • Very short delivery times for standard furnaces and spare parts
Additional equipment
  • Chimney, chimney with fan or catalytic converter
  • Over-temperature limit controller with adjustable cutout temperature for thermal protection
  • Protective gas connection to purge with non-flammable gases
  • Manual or automatic gas supply system
  • Port for thermocouple in the rear wall or in the furnace door
  • Process control and documentation via VCD software package for monitoring, documentation and control
Specifications
Specifications
1100°C
Furnace
Lift Door
50/60 Hz
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Nabertherm™ Muffle Furnace With Lift Door, 1100°C