missing translation for 'onlineSavingsMsg'
Learn More

Genlab Multi Purpose Benchtop Ovens

Highly efficient, reliable, and cost effective ovens that suit most warming, drying, and heat treatment processes. Available in a variety of sizes and equipped with intuitive touchscreen control that offers excellent accuracies.

£1640.00 - £2770.00

Specifications

Chamber Material Stainless Steel
Controller Type Base Mounted, Touchscreen
Description Multi-purpose benchtop oven with base mounted controller, fan assisted
Temperature (Metric) 40°C to 250°C
Material Coated Steel
View More Specs

Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
16305598
View Documents
Genlab
MINO/30/TDIG/F
30 L
£2770.00
Each
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
 
16315598
View Documents
Genlab
MINO/40/TDIG/F
40 L N/A N/A N/A
16325598
View Documents
Genlab
MINO/50/TDIG/F
53 L
£2275.00
Each
Estimated Shipment: 22-07-2025
to see stock.
Add to basket
 
16335598
View Documents
Genlab
MINO/75/TDIG/F
73 L
£2245.00
Each
Estimated Shipment: 22-07-2025
to see stock.
Add to basket
 
Description

Description

Included Features

  • On screen historical trending (48 Hours)
  • Manual or automatic overheat reset
  • High accuracy Pt100B duplex sensors
  • Excellent stability (fan assisted)
  • Low chamber uniformity
  • Energy efficient with integral energy meter
  • New - Set point lock feature

Available Options (Upon Request)

  • Fan assisted circulation (Not available < 30L)
  • Stainless steel chamber
  • 8 Stage profile control
  • Count down timer with 24/7 real time clock operation
  • 10 Menu x 8 Stage Profile Controller with Cyclic feature
  • Access ports (25, 50, 75 or 100 mm)
  • Audible warnings
  • Traceable calibration certificate to National Standards
  • Bespoke stands and stacking kits
  • Extractor unit
  • Extended warranty
  • Explosion relief panels (OV style only)
  • Bespoke solutions available upon request
Specifications

Specifications

Stainless Steel
Multi-purpose benchtop oven with base mounted controller, fan assisted
Coated Steel
Chrome Plated Wire Grid Shelves
Base Mounted, Touchscreen
40°C to 250°C
2 Shelves, 3 Positions
<±0.6°C (Fan Assisted)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Genlab Multi Purpose Benchtop Ovens

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.