Learn More
Hanna Instruments™ Multi-range EC Portable Meter
Brand:  Hanna Instruments™ HI9033
Product Code. 10517712
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Rugged, waterproof and simple to use with recessed rubber keyboard, large displays and automatic temperature compensation. Easy to clean sensor. Extremely robust, highly accurate potentiometric system of measurement. If you need to measure conductivity only, then the HI-9033 is ideal.
  • Simple to use, waterproof meter with single point calibration
  • Single probe measures from deionised water to brine over 4 different ranges
  • Automatic temperature compensation
  • Supplied with probe on 1m cable and carrying case
Specifications
Specifications
0.00 to 19.99 mS/cm
0.01 mS/cm
±1% Full-scale
1 μS/cm
Multi-range EC Portable Meter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products