Learn More
Thermo Scientific™ Multidrop™ Combi nL Reagent Dispenser, 50nL to 50μL

Thermo Scientific™ Multidrop™ Combi nL Rreagent Dispenser provides accurate, consistent and high throughput dispensing for all laboratories across a volume range of 50nL to 50μL.
Brand: Thermo Scientific™ 5840400
Description
Experience accurate, consistent, high throughput dispensing for all laboratories with the Thermo Scientific™ Multidrop™ Combi nL Reagent Dispenser. The Combi nL is a nano- to microvolume bulk reagent dispenser with a volume range of 50nL to 50μL. It expands the range of the market leading Multidrop product family, offering easy entry to precise, accurate and reliable low-volume dispensing in pharmaceutical and biotechnology laboratories.
Key Features:- Offers low volume dispensing for volumes 50nL -50μL into 96-, 384- and 1536-well plates
- Results in high quality assay data
- Increased flexibility—dispensing repeatably into plates with variable height allowing the user to fill either rows, columns, or wells with the easy to use FILLit Software
- Effortless dispensing—the visual, icon-based onboard user interface makes all functions easy to set tup, use, and maintain
- Reduced reagent costs—low volume dispensing, a minimal dead volume and the backflush function minimize the use of expensive reagents
- High throughput—fast dispensing combined with full robotic compatibility ensures increased assay throughput for laboratories needing low volume assay formats
Recommended for:
Assay development; Primary and secondary screening; Genomics and proteomics research; PCR set-up; Sequencing set-up; Cell based assays

Specifications
<1μL: ±5% >1μL: ±2% |
|
Multidrop Combi nL low volume reagent dispenser | |
1 nL (50 to 999 nL), 10 nL (1.00 to 9.99 μL), 100 nL (10.0 to 50.0 μL), 1 nL (FILLit Software) | |
96-, 384- and -1536-well plates | |
50 nL in 6 sec., 1 μL in 8 sec. (384-well plate) and 50 nL in 21 sec., 1 μL in 27 sec. (1536-well plate) | |
14 in. | |
8.6 in. | |
14.8 in. | |
21.2 lb. |
Reagent Dispenser | |
50 nL to 50 μL | |
Serial RS-232, USB | |
50nL: CV≤10%, 0.5μL:CV≤5%, 1-10μL:CV≤4%, >10μL: CV≤2% | |
<1.2 mL | |
35.5 cm | |
22 cm | |
37.5 cm | |
9.6 kg |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
838-8222 | Certificate of Compliance | 28/01/2025 | 5840400 |
838-8221 | Certificate of Compliance | 28/01/2025 | 5840400 |
838-8223 | Certificate of Compliance | 28/01/2025 | 5840400 |
838-8224 | Certificate of Compliance | 28/01/2025 | 5840400 |
838-8227 | Certificate of Compliance | 28/01/2025 | 5840400 |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Your input is important to us. Please complete this form to provide feedback related to the content on this product.