Learn More
Thermo Scientific™ Multidrop™ Pico 1 & Pico 8 Digital Dispenser
Miniaturize assays and achieve accurate results on picolter range with non-contact dispensing with eay to use software
Brand: Thermo Scientific™ 5840500
10913.00 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
Pico 1 and Pico 8 offers enhanced dispensing precision with volume as low as 11pL in any well, using up to ten times less reagent compared to manual pipetting workflows, enabling significant cost and waste reductions. Designed to enable assay miniaturization, boost productivity and improve accuracy of results makes them ideal for a broad array of low-volume applications such as qPCR, dose response curves, drug screening assays and ELISA.
Improved Efficiency and SpeedThe Digital Dispenser offers 40 times greater speed than analog serial dilution – from 10 minutes to just 15 seconds – eliminating precious time and accelerating your research.
Features:
- Cost Saving through assay miniaturization
- Improved efficiency and sample integrity
- Powerful, flexible, and easy to use software
The Digital Dispenser supports a wide range of applications and sample types:
- qPCR
- Drug combination studies
- DMPK
- Assay development
- Biochemical assays
- Secondary screening
- Cell-based assays
- Spotting
- Enzyme profiling
- Synthetic biology
- Antibody therapies
- SAR
The Digital Dispenser offers 40 times greater speed than analog serial dilution – from 10 minutes to just 15 seconds – eliminating precious time and accelerating your research.
Service Plan
Yes
Specifications
Digital Dispenser | |
11 pL to 20 μL, 1 nL to 200 μL | |
USB | |
≤ 8% | |
11 pL to 10 μL in seconds | |
5.9 | |
100/240 V |
Multidrop™ Pico 1 Digital Dispenser | |
Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting | |
12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm | |
PicoIT 1 | |
31 x 28 x 20 cm | |
100 to 240V 50/60Hz | |
50/60 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.