Learn More
WTW™ MultiLine™ Multi 3620 IDS Multi-parameter Portable Meter
Solve multiple measuring tasks at once and save time and costs. WTW™ MultiLine™ Multi 3620 IDS Multi-parameter Portable Meters feature two universal measuring channels for measuring pH, ORP, dissolved oxygen and/or turbidity simultaneously.
Brand:  WTW™ 2FD56G
 View more versions of this product
Product Code. 15520476
  • £2865.00 / Each
Explore more special offers
Description
Description
  • Wireless ready
  • Simultaneous display of all measured values
  • Digital sensor recognition
  • Two galvanically isolated measuring inputs
  • Dual – 2 channels, 4 parameters
  • Galvanic isolation of the sensor inputs prevents interferences of measuring signals
  • Use the new wireless modules together with new IDS plug head sensors, be independent from cables and measure even at hardly accessible locations
  • Efficient battery or mains supply, you will be able to measure cost-effectively in the field and in the lab
  • Data transfer either takes place via USB stick or via an USB connection directly to a computer
  • Color graphic display offers measured values as well as additional information such as sensor position, CMC and QSC
  • Traceability of the individual measured values can be supported by activating the user administration
Scope of delivery includes the MultiLab™ Importer to import data via Excel™ as well as the MultiLab™ User (user administration).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2
IP 67
Manual 500 / 10000 automatic
Wide range power supply, 4x 1.2 V NiMH battery or via Mini USB-B
LCD Color Graphic, Backlit
Set, SenTix 940-3, FDO 925-3, TetraCon 925-3
MultiLine™ Multi 3620 IDS Multi-parameter Portable Meter, Set G