Learn More
Fisherbrand™ multiSUB™ Midi Horizontal Gel System
Designed for routine horizontal gel electrophoresis.
Brand: Fisherbrand™ FMSMIDI7
This item is not returnable.
View return policy
Description
- Suitable for rapid electrophoresis
- Low buffer volumes
- Runs about 50 to 100 samples
Specifications
Horizontal Gel Electrophoresis System | |
multiSUB™ Midi electrophoresis unit, 10 x 7cm UV Tray, 2 x 16 sample combs, loading guides and dams | |
0.07 gal. | |
4.92 in. | |
8.66 in. | |
3.54 in. |
W x L: 10 x 7 cm (3.93 x 2.75 in.) | |
50 | |
300 mL | |
12.5 cm | |
22 cm | |
9 cm |