Learn More
MP Biomedicals™ Multitest Slide
Multitest Slide with a hydrophobic blue PTFE coating to keep your samples in the wells. Wells spacing is same as standard microplates. A 4-channel pipette can be used to transfer samples directly from the microplate to the slide. Appropriate for one-time use.
Brand:  MP Biomedicals™ 096041205E
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11411814
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The uncoated areas have a total wettable surface so that liquid spreads evenly over the area of each well. For cell attachment and growth, the slides require only a simple washing in deionized water or detergent followed by sterilization.
  • Self-draining surface
  • Sterilizable
  • Resistant to direct heat
  • Resistant to common organic solvents and fixatives
  • One-time use
  • Numbered
Specifications
Specifications
Cell Culture Slide
2 mm
7 mm
12
6 mm
100/Box
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.