Learn More
MP Biomedicals™ Multitest Slide
Multitest Slide with a hydrophobic blue PTFE coating to keep your samples in the wells. Wells spacing is same as standard microplates. A 4-channel pipette can be used to transfer samples directly from the microplate to the slide. Appropriate for one-time use.
Brand: MP Biomedicals™ 096041505E
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The uncoated areas have a total wettable surface so that liquid spreads evenly over the area of each well. For cell attachment and growth, the slides require only a simple washing in deionized water or detergent followed by sterilization.
- Self-draining surface
- Sterilizable
- Resistant to direct heat
- Resistant to common organic solvents and fixatives
- One-time use
- Numbered
Specifications
Cell Culture Slide | |
3 mm | |
4 mm |
15 | |
5 mm | |
100/Box |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.