Learn More
MP Biomedicals™ Multitest Slide
Multitest Slide with a hydrophobic blue PTFE coating to keep your samples in the wells. Wells spacing is same as standard microplates. A 4-channel pipette can be used to transfer samples directly from the microplate to the slide. Appropriate for one-time use.
Brand:  MP Biomedicals™ 096040805E
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11471804
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
The uncoated areas have a total wettable surface so that liquid spreads evenly over the area of each well. For cell attachment and growth, the slides require only a simple washing in deionized water or detergent followed by sterilization.
  • Self-draining surface
  • Sterilizable
  • Resistant to direct heat
  • Resistant to common organic solvents and fixatives
  • One-time use
  • Numbered
Specifications
Specifications
Cell Culture Slide
2 mm
6 mm
8
4 mm
100/Box
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.