missing translation for 'onlineSavingsMsg'
Learn More

ZVG™ Multizell™ Tissues

These tissue paper wipes are highly absorbent, lint-free, and made from 100% recycled material. The towels are wet-strength and environmentally friendly.

Brand:  ZVG™ 10440-00

Additional Details : Weight : 4.35000kg

 View more versions of this product

Product Code. 17873535

  • £54.94 / Pack of 1000
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

  • Ideal choice for when something needs to be quickly wiped up or cleaned
  • Absorbent and lint-free tissues can effectively remove both dry and oily dirt
  • Cost-effective Multizell™ tissues are made from 100% recycled material
  • Ideal for gas stations, car workshops, and even in the healthcare sector - whether it's for elderly and infant care or for cleaning soiled beds
  • Lint-free, highly absorbent, and highly resistant to moisture
  • Convenient single-sheet dispensing makes the environmentally friendly Multizell™ tissues significantly more economical to use compared to conventional layered pulp

In the zetPutz™ and zetMedica™ range, you will find more lint-free, highly absorbent 1- to 4-ply cleaning and polishing tissues for individual cleaning purposes.

Specifications

Specifications

Wiping Paper
27 cm
32 cm
Natural
3-ply
10.6 in.
12.6 in.
Recycled Paper
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers