missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MW Series Utility Cart with Wire Shelves
Utility carts are constructed of Super Erecta™ shelves with plastic split sleeves, two one-piece handles ,and casters with donut bumpers to make mobility of lab equipment easy.
Brand: Metro™ MW712
This item is not returnable.
View return policy
Description
- Cart load capacity 375 lb. (170.1kg)
- Fully interchangeable Super Erecta shelves
- Center shelf can be positioned at any height in 1 in. (2.5 cm) increments
- One-piece stainless-steel handles add rigidity and durability
- 18 in. (46 cm) carts have 4 in. dia. (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. dia. (13 cm) casters
- Three tiers
- Solid Shelves
- 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
- Raised edges help contain spills and make cleanup easier
- Sturdy corner castings facilitate mounting
Order Info
Shipped knocked down. Assembles/disassembles quickly and easily without tools.
Specifications
Utility Cart | |
NSF | |
3-tier Model | |
Transporting Materials | |
39 in. | |
Super Erecta™ 3 Stainless Steel wire shelves with plastic split sleeves, 2 one-piece Stainless Steel handle, 5 in. dia. caster, 3.5 in. donut bumpers | |
91 cm | |
170 kg | |
3 | |
24 in. |
170 kg | |
Silver | |
36 x 24 x 39 in. | |
Stainless Steel | |
99 cm | |
36 in. | |
375 lb. | |
Stainless Steel | |
Stainless Steel | |
61 cm |