missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MW Series Utility Cart with Wire Shelves

Utility carts are constructed of Super Erecta™ shelves with plastic split sleeves, two one-piece handles ,and casters with donut bumpers to make mobility of lab equipment easy.

£476.00 - £2135.00

Specifications

Product Type Utility Cart
Capacity (Metric) 170 kg
Certifications/Compliance NSF
Color Silver
Description 3-tier Model
View More Specs

Products 5
Product Code Brand Dimensions (L x W x H) Handle Material Shelf Material Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Handle Material Shelf Material Price Quantity & Availability  
15937181
View Documents
Metro™
MW712
36 x 24 x 39 in. Stainless Steel Stainless Steel
£2135.00
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
15997171
View Documents
Metro™
MW701
24 x 18 x 38 in. Chrome Chrome
£476.00
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
15927181
View Documents
Metro™
MW708
36 x 21 x 39 in. Stainless Steel Stainless Steel
£1475.00
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
15917181
View Documents
Metro™
MW704
30 x 18 x 38 in. Stainless Steel Stainless Steel
£1310.00
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
15907181
View Documents
Metro™
MW702
24 x 18 x 38 in. Stainless Steel Stainless Steel
£1190.00
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
Description

Description

  • Cart load capacity 375 lb. (170.1kg)
  • Fully interchangeable Super Erecta shelves
  • Center shelf can be positioned at any height in 1 in. (2.5 cm) increments
  • One-piece stainless-steel handles add rigidity and durability
  • 18 in. (46 cm) carts have 4 in. dia. (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. dia. (13 cm) casters
  • Three tiers
  • Solid Shelves
  • 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
  • Raised edges help contain spills and make cleanup easier
  • Sturdy corner castings facilitate mounting

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Specifications

Utility Cart
NSF
3-tier Model
375 lb.
3
170 kg
Silver
Transporting Materials
170 kg
Videos
SDS
Documents

Documents

Product Certifications