missing translation for 'onlineSavingsMsg'
Learn More

MX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13750-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16862124

  • £130.00 / 20µL
Estimated Shipment: 01-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody can recognize the endogenous Mx1 protein in several tissues like lung and spleen.

This gene encodes a guanosine triphosphate (GTP)-metabolizing protein that participates in the cellular antiviral response. The encoded protein is induced by type I and type II interferons and antagonizes the replication process of several different RNA and DNA viruses. There is a related gene located adjacent to this gene on chromosome 21, and there are multiple pseudogenes located in a cluster on chromosome 4. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

MX1
Polyclonal
Unconjugated
MX1
IFI 78K, IFI78, Interferon induced protein p78, MX, MX1, MxA, Myxovirus resistance protein 1
Rabbit
Antigen Affinity Chromatography
RUO
17857, 4599
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09922, P20591
Mx1
MX1 Fusion Protein Ag4692
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers