missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | MX1 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16862124
|
Proteintech
13750-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16852124
|
Proteintech
13750-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody can recognize the endogenous Mx1 protein in several tissues like lung and spleen.
This gene encodes a guanosine triphosphate (GTP)-metabolizing protein that participates in the cellular antiviral response. The encoded protein is induced by type I and type II interferons and antagonizes the replication process of several different RNA and DNA viruses. There is a related gene located adjacent to this gene on chromosome 21, and there are multiple pseudogenes located in a cluster on chromosome 4. Alternative splicing results in multiple transcript variants.Specifications
MX1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P09922, P20591 | |
17857, 4599 | |
MX1 Fusion Protein Ag4692 | |
Primary | |
-20°C | |
MX1 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
IFI 78K, IFI78, Interferon induced protein p78, MX, MX1, MxA, Myxovirus resistance protein 1 | |
Mx1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |