missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5A1F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60229-1-IG-20UL
This item is not returnable.
View return policy
Description
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
Myosin Light Chain 2 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08733, P10916, P51667 | |
Myl2 | |
Myosin Light Chain 2 Fusion Protein Ag1356 | |
20 μL | |
Primary | |
Porcine, Mouse, Rat, Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
5A1F1 | |
Unconjugated | |
MYL2 | |
MLC 2, MLC 2v, MLC2, MLC2v, MYL2, Myosin Light Chain 2 | |
Mouse | |
Protein G | |
RUO | |
17906, 363925, 396690, 4633 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Myosin Light Chain 2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5A1F1, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction