missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)piperazine, 98.5%, Thermo Scientific Chemicals

£26.90 - £91.30

Chemical Identifiers

CAS 103-76-4
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00005970
InChI Key WFCSWCVEJLETKA-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
PubChem CID 7677
IUPAC Name 2-piperazin-1-ylethanol
SMILES C1CN(CCN1)CCO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10704061
View Documents
Thermo Scientific Acros
121191000
100 g Glass bottle
£26.90
100g
Please to purchase this item. Need a web account? Register with us today!
10498840
View Documents
Thermo Scientific Acros
121195000
500 g Glass bottle
£91.30
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103-76-4
130.19
WFCSWCVEJLETKA-UHFFFAOYSA-N
7677
C1CN(CCN1)CCO
C6H14N2O
MFCD00005970
n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
2-piperazin-1-ylethanol

Specifications

103-76-4
100.0
Colorless to Yellow
245°C to 246°C
Authentic
C6H14N2O
MFCD00005970
1.06
Solubility in water: miscible with water. Other solubilities: miscible with most common organic solvents
C1CN(CCN1)CCO
130.19
130.19
Oily Liquid
98.0
-38.5°C
1.0600g/mL
135°C
98% min. (GC)
1.5055 to 1.5075
23, II, 6
n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
WFCSWCVEJLETKA-UHFFFAOYSA-N
2-piperazin-1-ylethanol
7677
98.50%
N-(2-Hydroxyethyl)piperazine, 98.5%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-142-3

missing translation for 'rtecsNumber' : TL6825000

missing translation for 'tsca' : TSCA

Recommended Storage : Yellowing of the substance during storage possible

Product Identifier
  • N-(2-Hydroxyethyl)piperazine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only