Learn More
N-(2-Hydroxyethyl)pyrrolidine, 97%, Thermo Scientific Chemicals
£41.10 - £117.00
Chemical Identifiers
CAS | 2955-88-6 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.18 |
MDL Number | MFCD00003181 |
InChI Key | XBRDBODLCHKXHI-UHFFFAOYSA-N |
Synonym | 1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol |
PubChem CID | 76288 |
ChEBI | CHEBI:48293 |
IUPAC Name | 2-pyrrolidin-1-ylethanol |
SMILES | OCCN1CCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446828
|
Thermo Scientific Alfa Aesar
H60368.AC |
25 mL |
£41.10
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15456828
|
Thermo Scientific Alfa Aesar
H60368.AE |
100 mL |
£117.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
Solubility
Partly soluble in water. Soluble in alcohol, ether, benzene.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
2955-88-6 | |
115.18 | |
XBRDBODLCHKXHI-UHFFFAOYSA-N | |
76288 | |
2-pyrrolidin-1-ylethanol |
C6H13NO | |
MFCD00003181 | |
1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol | |
CHEBI:48293 | |
OCCN1CCCC1 |
Specifications
2955-88-6 | |
79°C to 81°C | |
C6H13NO | |
MFCD00003181 | |
102976 | |
Partly soluble in water. Soluble in alcohol,ether,benzene. | |
OCCN1CCCC1 | |
115.18 | |
CHEBI:48293 | |
97% |
0.985 | |
56°C (133°F) | |
1.471 | |
UN1993 | |
1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol | |
XBRDBODLCHKXHI-UHFFFAOYSA-N | |
2-pyrrolidin-1-ylethanol | |
76288 | |
115.17 | |
N-(2-Hydroxyethyl)pyrrolidine |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 220-976-3
missing translation for 'rtecsNumber' : UY1175000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only