missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3,5-Dichlorophenyl)thiourea, 98+%, Thermo Scientific™
£21.50 - £260.00
Chemical Identifiers
CAS | 107707-33-5 |
---|---|
Molecular Formula | C7H6Cl2N2S |
Molecular Weight (g/mol) | 221.099 |
MDL Number | MFCD00041168 |
InChI Key | BNJARXOEVLYKDN-UHFFFAOYSA-N |
Synonym | 1-3,5-dichlorophenyl thiourea, n-3,5-dichlorophenyl thiourea, 3,5-dichlorophenyl thiourea, 1-3,5-dichlorophenyl-2-thiourea, thiourea, 3,5-dichlorophenyl, amino 3,5-dichlorophenyl amino methane-1-thione, thiourea;, maybridge1_007644, acmc-1bnq2 |
PubChem CID | 2737539 |
IUPAC Name | (3,5-dichlorophenyl)thiourea |
SMILES | C1=C(C=C(C=C1Cl)Cl)NC(=S)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11457877
|
Thermo Scientific Alfa Aesar
A16558.03 |
1 g |
£21.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467877
|
Thermo Scientific Alfa Aesar
A16558.06 |
5 g |
£67.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477877
|
Thermo Scientific Alfa Aesar
A16558.14 |
25 g |
£260.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
107707-33-5 | |
221.099 | |
BNJARXOEVLYKDN-UHFFFAOYSA-N | |
2737539 | |
C1=C(C=C(C=C1Cl)Cl)NC(=S)N |
C7H6Cl2N2S | |
MFCD00041168 | |
1-3,5-dichlorophenyl thiourea, n-3,5-dichlorophenyl thiourea, 3,5-dichlorophenyl thiourea, 1-3,5-dichlorophenyl-2-thiourea, thiourea, 3,5-dichlorophenyl, amino 3,5-dichlorophenyl amino methane-1-thione, thiourea;, maybridge1_007644, acmc-1bnq2 | |
(3,5-dichlorophenyl)thiourea |
Specifications
107707-33-5 | |
C7H6Cl2N2S | |
UN2811 | |
1-3,5-dichlorophenyl thiourea, n-3,5-dichlorophenyl thiourea, 3,5-dichlorophenyl thiourea, 1-3,5-dichlorophenyl-2-thiourea, thiourea, 3,5-dichlorophenyl, amino 3,5-dichlorophenyl amino methane-1-thione, thiourea;, maybridge1_007644, acmc-1bnq2 | |
C1=C(C=C(C=C1Cl)Cl)NC(=S)N | |
221.099 | |
221.11 | |
N-(3,5-Dichlorophenyl)thiourea |
175°C to 179°C | |
MFCD00041168 | |
3257049 | |
BNJARXOEVLYKDN-UHFFFAOYSA-N | |
(3,5-dichlorophenyl)thiourea | |
2737539 | |
≥98% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures