missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[4-(Trifluoromethyl)phenyl]thiourea, 98%, Thermo Scientific Chemicals
£46.60 - £149.00
Chemical Identifiers
CAS | 1736-72-7 |
---|---|
Molecular Formula | C8H7F3N2S |
Molecular Weight (g/mol) | 220.21 |
MDL Number | MFCD00041189 |
InChI Key | OWTDDZMFRLUBQI-UHFFFAOYSA-N |
Synonym | 1-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl phenyl thiourea, 1-4-trifluoromethyl phenyl-2-thiourea, 4-trifluoromethyl phenylthiourea, n-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl-phenyl-thiourea, thiourea, 4-trifluoromethyl phenyl, amino 4-trifluoromethyl phenyl amino methane-1-thione |
PubChem CID | 2777727 |
IUPAC Name | [4-(trifluoromethyl)phenyl]thiourea |
SMILES | NC(=S)NC1=CC=C(C=C1)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11398755
|
Thermo Scientific Alfa Aesar
L12355.03 |
1 g |
£46.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11308765
|
Thermo Scientific Alfa Aesar
L12355.06 |
5 g |
£149.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1736-72-7 | |
220.21 | |
OWTDDZMFRLUBQI-UHFFFAOYSA-N | |
2777727 | |
NC(=S)NC1=CC=C(C=C1)C(F)(F)F |
C8H7F3N2S | |
MFCD00041189 | |
1-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl phenyl thiourea, 1-4-trifluoromethyl phenyl-2-thiourea, 4-trifluoromethyl phenylthiourea, n-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl-phenyl-thiourea, thiourea, 4-trifluoromethyl phenyl, amino 4-trifluoromethyl phenyl amino methane-1-thione | |
[4-(trifluoromethyl)phenyl]thiourea |
Specifications
1736-72-7 | |
Odorless | |
MFCD00041189 | |
2695479 | |
OWTDDZMFRLUBQI-UHFFFAOYSA-N | |
[4-(trifluoromethyl)phenyl]thiourea | |
2777727 | |
98% |
142°C to 146°C | |
C8H7F3N2S | |
UN2811 | |
1-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl phenyl thiourea, 1-4-trifluoromethyl phenyl-2-thiourea, 4-trifluoromethyl phenylthiourea, n-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl-phenyl-thiourea, thiourea, 4-trifluoromethyl phenyl, amino 4-trifluoromethyl phenyl amino methane-1-thione | |
NC(=S)NC1=CC=C(C=C1)C(F)(F)F | |
220.21 | |
220.22 | |
N-[4-(Trifluoromethyl)phenyl]thiourea |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only