missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals
£10.15 - £198.00
Chemical Identifiers
CAS | 5891-45-2 |
---|---|
Molecular Formula | C17H23NO6 |
Molecular Weight (g/mol) | 337.37 |
MDL Number | MFCD00077016 |
InChI Key | VJECGKAFPHEJQS-UHFFFAOYNA-N |
Synonym | z-glu-otbu, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18089, 1-tert-butyl n-cbz-l-glutamate, l-glutamic acid, n-phenylmethoxy carbonyl-, 1-1,1-dimethylethyl ester, z-l-glutamic acid, a-tert.butyl ester |
PubChem CID | 6994036 |
IUPAC Name | 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid |
SMILES | CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 |
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
5891-45-2 | |
337.37 | |
VJECGKAFPHEJQS-UHFFFAOYNA-N | |
6994036 | |
CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 |
C17H23NO6 | |
MFCD00077016 | |
z-glu-otbu, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18089, 1-tert-butyl n-cbz-l-glutamate, l-glutamic acid, n-phenylmethoxy carbonyl-, 1-1,1-dimethylethyl ester, z-l-glutamic acid, a-tert.butyl ester | |
4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid |
Spezifikation
5891-45-2 | |
MFCD00077016 | |
VJECGKAFPHEJQS-UHFFFAOYNA-N | |
4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid | |
6994036 | |
95% | |
N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester |
C17H23NO6 | |
z-glu-otbu, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18089, 1-tert-butyl n-cbz-l-glutamate, l-glutamic acid, n-phenylmethoxy carbonyl-, 1-1,1-dimethylethyl ester, z-l-glutamic acid, a-tert.butyl ester | |
CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 | |
337.37 | |
337.37 | |
Solid |
Sicherheit und Handhabung
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only