missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-phenylalanine methyl ester, 95%, Thermo Scientific Chemicals
£81.20 - £241.00
Chemical Identifiers
CAS | 51987-73-6 |
---|---|
Molecular Formula | C15H21NO4 |
Molecular Weight (g/mol) | 279.34 |
MDL Number | MFCD00076977 |
InChI Key | SDSWSVBXRBXPRL-UHFFFAOYNA-N |
Synonym | boc-phe-ome, boc-l-phenylalanine methyl ester, l-boc-phenyl-alanine methyl ester, n-boc-l-phenylalanine methyl ester, n-tert-butoxycarbonyl-l-phenylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, s-2-tert-butoxycarbonylamino-3-phenyl-propionic acid methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, t-boc-l-phe-ome, pubchem13507 |
PubChem CID | 9882138 |
IUPAC Name | methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate |
SMILES | COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15429568
|
Thermo Scientific Alfa Aesar
H63078.14 |
25 g |
£81.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15439568
|
Thermo Scientific Alfa Aesar
H63078.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51987-73-6 | |
279.34 | |
SDSWSVBXRBXPRL-UHFFFAOYNA-N | |
9882138 | |
COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C |
C15H21NO4 | |
MFCD00076977 | |
boc-phe-ome, boc-l-phenylalanine methyl ester, l-boc-phenyl-alanine methyl ester, n-boc-l-phenylalanine methyl ester, n-tert-butoxycarbonyl-l-phenylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, s-2-tert-butoxycarbonylamino-3-phenyl-propionic acid methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, t-boc-l-phe-ome, pubchem13507 | |
methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate |
Specifications
36°C to 40°C | |
White | |
MFCD00076977 | |
boc-phe-ome, boc-l-phenylalanine methyl ester, l-boc-phenyl-alanine methyl ester, n-boc-l-phenylalanine methyl ester, n-tert-butoxycarbonyl-l-phenylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, s-2-tert-butoxycarbonylamino-3-phenyl-propionic acid methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, t-boc-l-phe-ome, pubchem13507 | |
COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C | |
279.34 | |
379.33 | |
113°C (235°F) | |
N-Boc-L-phenylalanine methyl ester |
51987-73-6 | |
C15H21NO4 | |
3061910 | |
SDSWSVBXRBXPRL-UHFFFAOYNA-N | |
methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate | |
9882138 | |
95% | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only