missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-trans-4-hydroxy-L-proline benzyl ester, 95%, Thermo Scientific Chemicals
£33.05 - £340.00
Chemical Identifiers
CAS | 89813-47-8 |
---|---|
Molecular Formula | C17H23NO5 |
Molecular Weight (g/mol) | 321.373 |
MDL Number | MFCD00076979 |
InChI Key | BEIPCYKSYYZEJH-KGLIPLIRSA-N |
Synonym | boc-hyp-obzl, boc-o-benzyl-l-hydroxyproline, 2s,4r-2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 2-benzyl 1-tert-butyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, bochypobn, boc-hyp-obn, 2s,4r-2-benzyl 1-t-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate |
PubChem CID | 11902907 |
IUPAC Name | 2-O-benzyl 1-O-tert-butyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate |
SMILES | CC(C)(C)OC(=O)N1CC(CC1C(=O)OCC2=CC=CC=C2)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15447496
|
Thermo Scientific Alfa Aesar
H62495.MD |
250 mg | |||||||
15419018
|
Thermo Scientific Alfa Aesar
H62495.03 |
1 g |
£110.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15429018
|
Thermo Scientific Alfa Aesar
H62495.06 |
5 g |
£340.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
89813-47-8 | |
321.373 | |
BEIPCYKSYYZEJH-KGLIPLIRSA-N | |
11902907 | |
CC(C)(C)OC(=O)N1CC(CC1C(=O)OCC2=CC=CC=C2)O |
C17H23NO5 | |
MFCD00076979 | |
boc-hyp-obzl, boc-o-benzyl-l-hydroxyproline, 2s,4r-2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 2-benzyl 1-tert-butyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, bochypobn, boc-hyp-obn, 2s,4r-2-benzyl 1-t-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate | |
2-O-benzyl 1-O-tert-butyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate |
Specifications
89813-47-8 | |
C17H23NO5 | |
boc-hyp-obzl, boc-o-benzyl-l-hydroxyproline, 2s,4r-2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 2-benzyl 1-tert-butyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, bochypobn, boc-hyp-obn, 2s,4r-2-benzyl 1-t-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate | |
CC(C)(C)OC(=O)N1CC(CC1C(=O)OCC2=CC=CC=C2)O | |
321.373 | |
321.37 | |
Liquid |
Brown | |
MFCD00076979 | |
BEIPCYKSYYZEJH-KGLIPLIRSA-N | |
2-O-benzyl 1-O-tert-butyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate | |
11902907 | |
95% | |
N-Boc-trans-4-hydroxy-L-proline benzyl ester |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only