Learn More
n-Decylamine, 99%, Thermo Scientific Chemicals
£43.22 - £139.70
Chemical Identifiers
CAS | 2016-57-1 |
---|---|
Molecular Formula | C10H23N |
Molecular Weight (g/mol) | 157.30 |
MDL Number | MFCD00008149 |
InChI Key | MHZGKXUYDGKKIU-UHFFFAOYSA-N |
Synonym | decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine |
PubChem CID | 8916 |
IUPAC Name | decan-1-amine |
SMILES | CCCCCCCCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10008622
|
Thermo Scientific Acros
111920100 |
10 g | Glass bottle |
£43.22
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606624
|
Thermo Scientific Acros
111920500 |
50 g | Glass bottle |
£139.70
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2016-57-1 | |
157.30 | |
MHZGKXUYDGKKIU-UHFFFAOYSA-N | |
8916 | |
CCCCCCCCCCN |
C10H23N | |
MFCD00008149 | |
decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine | |
decan-1-amine |
Specifications
2016-57-1 | |
100.0 | |
Brown or Colorless to Yellow | |
216°C to 218°C | |
Authentic | |
C10H23N | |
1.4350 to 1.437 | |
04, 199 | |
decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine | |
CCCCCCCCCCN | |
157.30 | |
157.3 | |
Liquid |
98.5 | |
12°C to 15°C | |
0.7870g/mL | |
85°C | |
98.5% min. (GC) | |
CH3(CH2)9NH2 | |
MFCD00008149 | |
0.787 | |
MHZGKXUYDGKKIU-UHFFFAOYSA-N | |
decan-1-amine | |
8916 | |
99% | |
n-Decylamine |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT i
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-957-7
RUO – Research Use Only